(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl 98%

Supplier: Ambeed
Warning

A751046-100MG A751046-1G A751046-250MG
77255-602EA 220 USD
77255-602 77255-604 77255-606
(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl 98%
(R)-2,2'-Bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl

Precaución: for Research Use Only

Formula: C₇₆H₁₀₈O₈P₂
MDL Number: MFCD20265636
CAS Number: 1365531-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR