3-(Methylthio)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Thioanisoleboronic acid, 3-(Methylthio)benzeneboronic acid

A750657-10G A750657-25G A750657-5G A750657-1G
77281-132EA 32.96 USD
77281-132 77281-134 77281-136 77749-507
3-(Methylthio)phenylboronic acid 98%
3-(Methylthio)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₂S
MW: 168.02 g/mol
MDL Number: MFCD01319105
CAS Number: 128312-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR