3-Nitro-5-(trifluoromethyl)benzonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 3-Cyano-5-nitrobenzotrifluoride

A750024-100G A750024-1G A750024-25G A750024-5G
77281-124EA 444.96 USD
77281-124 77281-126 77281-128 77281-130
3-Nitro-5-(trifluoromethyl)benzonitrile 98%
3-Nitro-5-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃F₃N₂O₂
MW: 216.1200 g/mol
MDL Number: MFCD03412207
CAS Number: 20566-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR