2,2'-Bithiophene-5,5'-dicarboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: [2,2'-Bithiophene]-5,5'-dicarbaldehyde, 5,5'-Diformyl-2,2'-bithiophene

A749829-1G A749829-250MG A749829-5G
77255-588EA 24 USD
77255-588 77255-590 77255-592
2,2'-Bithiophene-5,5'-dicarboxaldehyde 98%
2,2'-Bithiophene-5,5'-dicarboxaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₆O₂S₂
MW: 222.28 g/mol
MDL Number: MFCD00839981
CAS Number: 32364-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR