5-(Dicyclohexylphosphino)-1',3',5'-triphenyl-[1,4']-bi-1H-pyrazole 98%

Supplier: Ambeed
Warning

A749452-1G A749452-100MG A749452-250MG A749452-50MG A749452-5G
77255-586EA 163 USD
77255-586 77618-062 77618-064 77618-066 77747-478
5-(Dicyclohexylphosphino)-1',3',5'-triphenyl-[1,4']-bi-1H-pyrazole 98%
5-(Dicyclohexylphosphino)-1',3',5'-triphenyl-[1,4']-bi-1H-pyrazole

Precaución: for Research Use Only

Formula: C₃₆H₃₉N₄P
MW: 558.69 g/mol
MDL Number: MFCD11616511
CAS Number: 1021176-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR