(4-Morpholinophenyl)methanamine hydrochloride 97%

Supplier: Ambeed
Warning

A748566-100MG A748566-1G A748566-250MG
77267-842EA 30 USD
77267-842 77267-844 77267-846
(4-Morpholinophenyl)methanamine hydrochloride 97%
(4-Morpholinophenyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₇ClN₂O
MDL Number: MFCD25955516
CAS Number: 1106986-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR