3-Amino-4,5-dimethylbenzoic acid 97%

Supplier: Ambeed
Warning

A748179-250MG A748179-1G
77267-838EA 78.28 USD
77267-838 77573-392
3-Amino-4,5-dimethylbenzoic acid 97%
3-Amino-4,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD17011765
CAS Number: 348165-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR