2-Fluoro-6-(trifluoromethyl)benzylamine 96%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-(Aminomethyl)-3-fluorobenzotrifluoride, [2-Fluoro-6-(trifluoromethyl)phenyl]methylamine
A747940-100G
A747940-1G
A747940-250MG
A747940-25G
A747940-5G
77267-824EA
243
USD
77267-824
77267-826
77267-828
77267-830
77267-832
77267-834
2-Fluoro-6-(trifluoromethyl)benzylamine 96%
2-Fluoro-6-(trifluoromethyl)benzylamine
Precaución: for Research Use Only
Formula:
C₈H₇F₄N |
MDL Number:
MFCD00061165 CAS Number: 239087-06-0 UN: 2735 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...