4-Chloro-2,6-xylenol 98%

Supplier: Ambeed
Warning

A747460-1G A747460-5G
77255-576EA 9 USD
77255-576 77255-578
4-Chloro-2,6-xylenol 98%
4-Chloro-2,6-xylenol

Precaución: for Research Use Only

Formula: C₈H₉ClO
MDL Number: MFCD00020138
CAS Number: 1123-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR