(S)-2-(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methylbutanamido)acetic acid 98% + (by HPLC)

Supplier: Ambeed
Danger

A746652-1G A746652-250MG A746652-5G
77267-816EA 115.36 USD
77267-816 77267-818 77267-820
(S)-2-(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methylbutanamido)acetic acid 98% + (by HPLC)
(S)-2-(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methylbutanamido)acetic acid

Precaución: for Research Use Only

Formula: C₂₂H₂₄N₂O₅
MW: 396.43 g/mol
MDL Number: MFCD18427309
CAS Number: 142810-19-3
UN: 3077
ADR: 9,Ⅲ

Order Now

Specification Test Results

Loss on drying < 5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR