2-(Methylthio)benzoic acid 98%

Supplier: Ambeed
Warning

A746023-1G A746023-25G A746023-5G
77255-570EA 9 USD
77255-570 77255-572 77255-574
2-(Methylthio)benzoic acid 98%
2-(Methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₈O₂S
MDL Number: MFCD00029934
CAS Number: 3724-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR