4-Fluorobenzyl mercaptan 98%

Supplier: Ambeed
Danger

A745533-1G A745533-25G A745533-5G
77293-510EA 21 USD
77293-510 77293-512 77293-514
4-Fluorobenzyl mercaptan 98%
4-Fluorobenzyl mercaptan

Precaución: for Research Use Only

Formula: C₇H₇FS
MDL Number: MFCD00040724
CAS Number: 15894-04-9
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR