2-Methyl-5-nitropyrimidine-4,6-diol 95%

Supplier: Ambeed
Warning

A745303-100MG A745303-1G A745303-250MG
77281-112EA 49 USD
77281-112 77281-114 77281-116
2-Methyl-5-nitropyrimidine-4,6-diol 95%
2-Methyl-5-nitropyrimidine-4,6-diol

Precaución: for Research Use Only

Formula: C₅H₅N₃O₄
MDL Number: MFCD00193996
CAS Number: 53925-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR