5-Methylthieno[2,3-d]pyrimidin-4(3H)-one 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methylthieno[2,3,d]pyrimidin-4(3H)-one

A745170-100MG A745170-250MG
77293-502EA 47.38 USD
77293-502 77293-504
5-Methylthieno[2,3-d]pyrimidin-4(3H)-one 98%
5-Methylthieno[2,3-d]pyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₇H₆N₂OS
MW: 166.2 g/mol
MDL Number: MFCD00205193
CAS Number: 43088-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR