4,4'-Azanediyldibenzoic acid 97%

Supplier: Ambeed
Warning

A744056-100MG A744056-250MG
77293-498EA 36.05 USD
77293-498 77293-500
4,4'-Azanediyldibenzoic acid 97%
4,4'-Azanediyldibenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO₄
MW: 257.24 g/mol
MDL Number: MFCD06408338
CAS Number: 20800-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR