1-Chloro-2,3-difluorobenzene 98%

Supplier: Ambeed
Danger

A743348-10G A743348-1G A743348-25G A743348-5G
77255-556EA 58.71 USD
77255-556 77255-558 77255-560 77255-562
1-Chloro-2,3-difluorobenzene 98%
1-Chloro-2,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClF₂
MW: 148.5400 g/mol
MDL Number: MFCD04112584
CAS Number: 36556-47-5
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR