4,6-Dichloro-2,5-diphenylpyrimidine 98%

Supplier: Ambeed
Warning

A743150-1G A743150-250MG A743150-25G
77293-488EA 45.32 USD
77293-488 77293-490 77293-492
4,6-Dichloro-2,5-diphenylpyrimidine 98%
4,6-Dichloro-2,5-diphenylpyrimidine

Precaución: for Research Use Only

Formula: C₁₆H₁₀Cl₂N₂
MW: 301.17 g/mol
MDL Number: MFCD00209572
CAS Number: 29133-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR