2,3-Diphenylbutane-2,3-diol 98+%

Supplier: Ambeed
Warning

A743074-1G A743074-250MG A743074-5G
77255-544EA 69.01 USD
77255-544 77255-546 77255-548
2,3-Diphenylbutane-2,3-diol 98+%
2,3-Diphenylbutane-2,3-diol

Precaución: for Research Use Only

Formula: C₁₆H₁₈O₂
MW: 242.31 g/mol
MDL Number: MFCD00004452
CAS Number: 1636-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR