2,3-Butanediol 98% mixture of stereoisomers

Supplier: Ambeed
Warning

A742684-100G A742684-25G A742684-500G A742684-5G
77255-536EA 15 USD
77255-536 77255-538 77255-540 77255-542
2,3-Butanediol 98% mixture of stereoisomers
2,3-Butanediol

Precaución: for Research Use Only

Formula: C₄H₁₀O₂
MDL Number: MFCD00004523
CAS Number: 513-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR