Aleuritic acid 95%

Supplier: Ambeed
Warning

A742536-100MG A742536-1G A742536-25G A742536-5G
77255-530EA 5 USD
77255-530 77255-532 77255-534 77618-042
Aleuritic acid 95%
Aleuritic acid

Precaución: for Research Use Only

Formula: C₁₆H₃₂O₅
MDL Number: MFCD00135596
CAS Number: 533-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR