SU-6656 98%

Supplier: Ambeed
Warning

Synonyms: N,N-Dimethyl-2-oxo-3-((4,5,6,7-tetrahydro-1H-indol-2-yl)methylene)indoline-5-sulphonamide, SU6656, (Z)-2-hydroxy-N,N-dimethyl-3-((4,5,6,7-tetrahydro-1H-indol-2-yl)methylene)-3H-indole-5-sulphonamide

A742025-10MG A742025-1MG A742025-50MG
77243-854EA 85 USD
77243-854 77243-856 77243-858
SU-6656 98%
SU-6656

Precaución: for Research Use Only

Formula: C₁₉H₂₁N₃O₃S
MDL Number: MFCD10565928
CAS Number: 330161-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR