N,N'-((9H-Fluorene-9,9-diyl)bis(4,1-phenylene))bis(N-([1,1'-biphenyl]-4-yl)-[1,1'-biphenyl]-4-amine) 97%

Supplier: Ambeed
Warning

A741592-100MG A741592-250MG
77255-526EA 40.17 USD
77255-526 77255-528
N,N'-((9H-Fluorene-9,9-diyl)bis(4,1-phenylene))bis(N-([1,1'-biphenyl]-4-yl)-[1,1'-biphenyl]-4-amine) 97%
N,N'-((9H-Fluorene-9,9-diyl)bis(4,1-phenylene))bis(N-([1,1'-biphenyl]-4-yl)-[1,1'-biphenyl]-4-amine)

Precaución: for Research Use Only

Formula: C₇₃H₅₂N₂
MW: 957.20 g/mol
MDL Number: MFCD12022430
CAS Number: 174141-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR