3-Methyl-1,1-diphenylurea 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-3,3-diphenylurea

A741526-100G A741526-1G A741526-25G A741526-5G
77281-088EA 229.69 USD
77281-088 77281-090 77281-092 77281-094
3-Methyl-1,1-diphenylurea 98%
3-Methyl-1,1-diphenylurea

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₂O
MW: 226.274 g/mol
MDL Number: MFCD00043722
CAS Number: 13114-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR