2-(2,3-Difluorophenyl)ethanol 95%

Supplier: Ambeed
Warning

A740059-1G A740059-250MG A740059-5G
77243-840EA 58.71 USD
77243-840 77243-842 77243-844
2-(2,3-Difluorophenyl)ethanol 95%
2-(2,3-Difluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₈F₂O
MW: 158.15 g/mol
MDL Number: MFCD01862011
CAS Number: 126163-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR