3-Nitrophenetole 99%

Supplier: Ambeed
Warning

Synonyms: 1-Ethoxy-3-nitrobenzene

A739918-10G A739918-1G A739918-25G A739918-5G
77293-480EA 52.53 USD
77293-480 77293-482 77293-484 77293-486
3-Nitrophenetole 99%
3-Nitrophenetole

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD00024472
CAS Number: 621-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR