1-Aminocyclobutanecarboxylic acid 95%

Supplier: Ambeed
Warning

A737321-100G A737321-10G A737321-1G A737321-25G A737321-5G
77281-068EA 347.11 USD
77281-068 77281-070 77281-072 77281-074 77281-076
1-Aminocyclobutanecarboxylic acid 95%
1-Aminocyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₉NO₂
MDL Number: MFCD00661068
CAS Number: 22264-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR