4-(Propan-2-yl)-1H-pyrazol-3-amine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Isopropyl-1H-pyrazol-5-amine

A736581-100MG A736581-1G A736581-250MG A736581-5G
77243-816EA 20.6 USD
77243-816 77243-818 77243-820 77618-028
4-(Propan-2-yl)-1H-pyrazol-3-amine 95%
4-(Propan-2-yl)-1H-pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₆H₁₁N₃
MW: 125.172 g/mol
MDL Number: MFCD11112127
CAS Number: 151521-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR