4,5-Difluoro-2-methoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A736177-1G A736177-250MG
77243-812EA 26 USD
77243-812 77243-814
4,5-Difluoro-2-methoxybenzaldehyde 98%
4,5-Difluoro-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MDL Number: MFCD06246884
CAS Number: 145742-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR