3-Benzo[b]thiophenesulfonyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: 1-Benzothiophene-3-sulphonyl Chloride, Benzothiophene-3-sulphonyl chloride

A736031-100MG A736031-1G A736031-250MG
77243-806EA 22.66 USD
77243-806 77243-808 77243-810
3-Benzo[b]thiophenesulfonyl chloride 95%
3-Benzo[b]thiophenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₈H₅ClO₂S₂
MW: 232.71 g/mol
MDL Number: MFCD02681897
CAS Number: 18494-87-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR