4-(Chloromethyl)benzaldehyde 95%

Supplier: Ambeed
Danger

A735505-10G A735505-1G A735505-250MG A735505-25G A735505-5G
77293-468EA 57 USD
77293-468 77293-470 77293-472 77293-474 77293-476
4-(Chloromethyl)benzaldehyde 95%
4-(Chloromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇ClO
MDL Number: MFCD00191396
CAS Number: 73291-09-5
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR