Ethyl-2-cyano-2-methylpropionate 98%
Supplier: Ambeed
|
Warning
|
A735493-100G
A735493-25G
A735493-500G
A735493-5G
77267-780EA
158
USD
77267-780
77267-782
77267-784
77267-786
Ethyl-2-cyano-2-methylpropionate 98%
Ethyl-2-cyano-2-methylpropionate
Precaución: for Research Use Only
Formula:
C₇H₁₁NO₂ |
MDL Number:
MFCD00034667 CAS Number: 1572-98-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...