4-(4-Chlorophenoxy)benzoic acid 98%
Supplier: Ambeed
|
Danger
|
A735464-100MG
A735464-1G
A735464-250MG
A735464-5G
77293-460EA
11
USD
77293-460
77293-462
77293-464
77293-466
4-(4-Chlorophenoxy)benzoic acid 98%
4-(4-Chlorophenoxy)benzoic acid
Precaución: for Research Use Only
Formula:
C₁₃H₉ClO₃ |
MDL Number:
MFCD01862494 CAS Number: 21120-67-2 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...