(E)-3-(4-Fluorophenyl)acrylaldehyde 95%

Supplier: Ambeed
Danger

A735410-10G A735410-25G A735410-5G A735410-1G
77267-774EA 42 USD
77267-774 77267-776 77267-778 77618-016
(E)-3-(4-Fluorophenyl)acrylaldehyde 95%
(E)-3-(4-Fluorophenyl)acrylaldehyde

Precaución: for Research Use Only

Formula: C₉H₇FO
MDL Number: MFCD03425895
CAS Number: 51791-26-5
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR