tert-Butyl 1-bromo-3,6,9,12-tetraoxapentadecan-15-oate 98%

Supplier: Ambeed
Warning

Synonyms: Br-PEG4-C2-Boc

A735322-1G A735322-5G A735322-100MG A735322-250MG
77267-772EA 268.83 USD
77267-772 77618-010 77749-492 77749-493
tert-Butyl 1-bromo-3,6,9,12-tetraoxapentadecan-15-oate 98%
tert-Butyl 1-bromo-3,6,9,12-tetraoxapentadecan-15-oate

Precaución: for Research Use Only

Formula: C₁₅H₂₉BrO₆
MW: 385.29 g/mol
MDL Number: MFCD22574808
CAS Number: 564476-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR