2,2',5,5'-Tetrachlorobenzidine 98%
Supplier: Ambeed
|
Warning
|
A734418-100G
A734418-25G
77255-494EA
21
USD
77255-494
77255-496
2,2',5,5'-Tetrachlorobenzidine 98%
2,2',5,5'-Tetrachlorobenzidine
Precaución: for Research Use Only
Formula:
C₁₂H₈Cl₄N₂ |
MDL Number:
MFCD00043918 CAS Number: 15721-02-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...