(1S,2S)-N,N'-Bis(2-acetyl-3-oxo-2-butenylidene)-1,2-dimesitylethylenediaminato cobalt(II) 98%

Supplier: Ambeed
Warning

Synonyms: (S)-AMAC

A734056-100MG A734056-250MG
77255-490EA 121 USD
77255-490 77255-492
(1S,2S)-N,N'-Bis(2-acetyl-3-oxo-2-butenylidene)-1,2-dimesitylethylenediaminato cobalt(II) 98%
(1S,2S)-N,N'-Bis(2-acetyl-3-oxo-2-butenylidene)-1,2-dimesitylethylenediaminato cobalt(II)

Precaución: for Research Use Only

Formula: C₃₂H₃₈CoN₂O₄
MW: 573.60 g/mol
MDL Number: MFCD06797062
CAS Number: 259259-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR