2-Ethyl-5,7-dimethyl-1H-imidazo[4,5-b]pyridine 98%

Supplier: Ambeed
Warning

Synonyms: Adenosine receptor antagonist 4

A733869-100MG A733869-250MG A733869-1G
77243-798EA 14 USD
77243-798 77243-800 77573-362
2-Ethyl-5,7-dimethyl-1H-imidazo[4,5-b]pyridine 98%
2-Ethyl-5,7-dimethyl-1H-imidazo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₃N₃
MDL Number: MFCD17779284
CAS Number: 133240-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR