4-(4-Iodophenyl)morpholine 98%
Supplier: Ambeed
|
Warning
|
A733181-1G
A733181-250MG
A733181-25G
77293-454EA
32.96
USD
77293-454
77293-456
77293-458
4-(4-Iodophenyl)morpholine 98%
4-(4-Iodophenyl)morpholine
Precaución: for Research Use Only
Formula:
C₁₀H₁₂INO |
MDL Number:
MFCD04974040 CAS Number: 87350-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...