N-Boc-PEG6-alcohol 98%

Supplier: Ambeed
Warning

A732776-1G A732776-250MG A732776-100MG
77267-758EA 79 USD
77267-758 77267-760 77617-982
N-Boc-PEG6-alcohol 98%
N-Boc-PEG6-alcohol

Precaución: for Research Use Only

Formula: C₁₇H₃₅NO₈
MDL Number: MFCD28385472
CAS Number: 331242-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR