Acetonylacetone 98%

Supplier: Ambeed
Warning

A732595-100G A732595-10G A732595-25G A732595-500G
77255-480EA 20 USD
77255-480 77255-482 77255-484 77255-486
Acetonylacetone 98%
Acetonylacetone

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MDL Number: MFCD00008792
CAS Number: 110-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR