5-Hydroxy-1H-indazole 98%

Supplier: Ambeed
Warning

Synonyms: 1H-Indazol-5-ol

A732478-100G A732478-10G A732478-1G A732478-250MG A732478-25G A732478-500G A732478-5G
77243-782EA 225.57 USD
77243-782 77243-784 77243-786 77243-788 77243-790 77243-792 77243-794
5-Hydroxy-1H-indazole 98%
5-Hydroxy-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.14 g/mol
MDL Number: MFCD01938124
CAS Number: 15579-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR