2',3',5'-Trifluoroacetophenone 96%

Supplier: Ambeed
Warning

A732198-1G A732198-25G A732198-5G
77255-474EA 20 USD
77255-474 77255-476 77255-478
2',3',5'-Trifluoroacetophenone 96%
2',3',5'-Trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅F₃O
MDL Number: MFCD00083548
CAS Number: 243459-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR