3-Chloro-2,5,6-trifluoropyridine 98%

Supplier: Ambeed
Danger

A732010-10G A732010-1G A732010-5G A732010-25G
77281-038EA 86.52 USD
77281-038 77281-040 77281-042 77749-485
3-Chloro-2,5,6-trifluoropyridine 98%
3-Chloro-2,5,6-trifluoropyridine

Precaución: for Research Use Only

Formula: C₅HClF₃N
MDL Number: MFCD03001160
CAS Number: 2879-42-7
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR