(R)-3-(Methylsulfonyl)piperidine hydrochloride 97%

Supplier: Ambeed
Warning

A730394-1G A730394-250MG
77255-460EA 320 USD
77255-460 77255-462
(R)-3-(Methylsulfonyl)piperidine hydrochloride 97%
(R)-3-(Methylsulfonyl)piperidine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO₂S
MDL Number: MFCD29918530
CAS Number: 1946010-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR