Sodium 3-nitrobenzenesulfonate 98%

Supplier: Ambeed
Warning

A730105-1000G A730105-100G A730105-25G A730105-500G
77293-440EA 27 USD
77293-440 77293-442 77293-444 77293-446
Sodium 3-nitrobenzenesulfonate 98%
Sodium-3-nitrobenzenesulphonate

Precaución: for Research Use Only

Formula: C₆H₄NNaO₅S
MDL Number: MFCD00007490
CAS Number: 127-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR