(2S)-1-[(4R)-4,5-dihydro-4-phenyl-2-oxazolyl]-2-(diphenylphosphino)ferrocene ≥98%, ee 99%

Supplier: Ambeed
Warning

A730002-100MG A730002-1G A730002-250MG A730002-5G
77255-456EA 24 USD
77255-456 77255-458 77573-342 77747-442
(2S)-1-[(4R)-4,5-dihydro-4-phenyl-2-oxazolyl]-2-(diphenylphosphino)ferrocene ≥98%, ee 99%
(2S)-1-[(4R)-4,5-dihydro-4-phenyl-2-oxazolyl]-2-(diphenylphosphino)ferrocene

Precaución: for Research Use Only

Formula: C₃₁H₂₆FeNOP
MW: 515.4 g/mol
MDL Number: MFCD30475631
CAS Number: 291536-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR