4,7,10,13,16,19,22,25,28-Nonaoxahentriacontane-1,31-dioic acid 97%

Supplier: Ambeed
Warning

A729927-1G A729927-250MG A729927-100MG
77267-744EA 174 USD
77267-744 77267-746 77749-484
4,7,10,13,16,19,22,25,28-Nonaoxahentriacontane-1,31-dioic acid 97%
4,7,10,13,16,19,22,25,28-Nonaoxahentriacontane-1,31-dioic acid

Precaución: for Research Use Only

Formula: C₂₂H₄₂O₁₃
MDL Number: MFCD11041130
CAS Number: 1268488-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR