2-Chloro-6-methylbenzaldehyde 98%

Supplier: Ambeed
Warning

A729754-1G A729754-250MG A729754-5G A729754-10G
77243-762EA 16 USD
77243-762 77243-764 77243-766 77747-440
2-Chloro-6-methylbenzaldehyde 98%
2-Chloro-6-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇ClO
MDL Number: MFCD01934409
CAS Number: 1194-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR