(R)-4-[2-(Diphenylphosphino)-1-naphthalenyl]-N-[(R)-1-phenylethyl]-1-phthalazinamine 98%

Supplier: Ambeed
Warning

A729275-100MG A729275-1G A729275-250MG
77255-438EA 75 USD
77255-438 77255-440 77255-442
(R)-4-[2-(Diphenylphosphino)-1-naphthalenyl]-N-[(R)-1-phenylethyl]-1-phthalazinamine 98%
(R)-4-[2-(Diphenylphosphino)-1-naphthalenyl]-N-[(R)-1-phenylethyl]-1-phthalazinamine

Precaución: for Research Use Only

Formula: C₃₈H₃₀N₃P
MDL Number: MFCD09264274
CAS Number: 828927-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR