Azobenzene-3,3'-dicarboxylic acid 95%

Supplier: Ambeed
Warning

A729222-1G A729222-250MG A729222-5G
77281-018EA 36 USD
77281-018 77281-020 77281-022
Azobenzene-3,3'-dicarboxylic acid 95%
Azobenzene-3,3'-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀N₂O₄
MDL Number: MFCD03844759
CAS Number: 621-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR